Hex-5-yn-1-amine hydrochloride 95%

Supplier: Ambeed
Warning

A524018-100MG A524018-1G A524018-250MG A524018-5G
76991-480EA 77.45 USD
76991-480 76991-482 76991-484 77684-357
Hex-5-yn-1-amine hydrochloride 95%
Hex-5-yn-1-amine hydrochloride

Precaución: For research use only.

Formula: C₆H₁₂ClN
MW: 133.61 g/mol
MDL Number: MFCD18839147
CAS Number: 102169-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR