4-(3-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-((3-Chlorobenzyl)oxy)-3,5-dimethylphenyl)boronic acid

A521911-1G A521911-250MG A521911-5G
77017-406EA 160.2 USD
77017-406 77017-408 77017-410
4-(3-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid 95%
4-(3-Chlorobenzyloxy)-3,5-dimethylphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₆BClO₃
MW: 290.5500 g/mol
MDL Number: MFCD06411362
CAS Number: 849062-21-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR