Boc-Trp(Boc)-OH 98%

Supplier: Ambeed
Warning

Synonyms: N-Boc-1-Boc-L-tryptophan, (S)-3-(1-(tert-Butoxycarbonyl)-1H-indol-3-yl)-2-((tert-butoxycarbonyl)amino)propanoic acid

A521732-1G A521732-5G A521732-25G A521732-10G
77004-854EA 10 USD
77004-854 77004-856 77602-232 77673-388
Boc-Trp(Boc)-OH 98%
Boc-Trp(Boc)-OH

Precaución: for Research Use Only

Formula: C₂₁H₂₈N₂O₆
MW: 404.463 g/mol
MDL Number: MFCD00270520
CAS Number: 144599-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR