4-Bromoindan-2-one 96%

Supplier: Ambeed
Warning

A521538-5G A521538-1G A521538-100MG A521538-250MG
77034-668EA 334 USD
77034-668 77572-656 77669-304 77872-875
4-Bromoindan-2-one 96%
4-Bromoindan-2-one

Precaución: for Research Use Only

Formula: C₉H₇BrO
MDL Number: MFCD09744095
CAS Number: 846032-36-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR