5-Cyclopropyl-1,3,4-thiadiazol-2-amine 95%

Supplier: Ambeed
Danger

A521394-1G A521394-250MG A521394-5G
77017-396EA 50.93 USD
77017-396 77017-398 77017-400
5-Cyclopropyl-1,3,4-thiadiazol-2-amine 95%
5-Cyclopropyl-1,3,4-thiadiazol-2-amine

Precaución: for Research Use Only

Formula: C₅H₇N₃S
MDL Number: MFCD00051649
CAS Number: 57235-50-4
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR