2-Cyano-N-(3-pyridinyl)acetamide 95%

Supplier: Ambeed
Warning

A521340-250MG A521340-5G
77004-842EA 28 USD
77004-842 77004-844
2-Cyano-N-(3-pyridinyl)acetamide 95%
2-Cyano-N-(3-pyridinyl)acetamide

Precaución: for Research Use Only

Formula: C₈H₇N₃O
MDL Number: MFCD00127642
CAS Number: 219618-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR