2-Acetylamino-5-bromo-4-methylpyridine 98%

Supplier: Ambeed
Warning

A520945-250MG A520945-25G A520945-5G
77004-836EA 16 USD
77004-836 77004-838 77004-840
2-Acetylamino-5-bromo-4-methylpyridine 98%
2-Acetylamino-5-bromo-4-methylpyridine

Precaución: for Research Use Only

Formula: C₈H₉BrN₂O
MDL Number: MFCD06798092
CAS Number: 142404-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR