Ethyl 2-chloro-4-methylbenzoate 95+%

Supplier: Ambeed
Warning

A520256-100G A520256-25G A520256-5G
77017-390EA 154.9 USD
77017-390 77017-392 77017-394
Ethyl 2-chloro-4-methylbenzoate 95+%
Ethyl 2-chloro-4-methylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO₂
MW: 198.64 g/mol
MDL Number: MFCD16250088
CAS Number: 99500-35-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR