2,3-Dibromotoluene 95%

Supplier: Ambeed
Warning

Synonyms: 1,2-Dibromo-3-methylbenzene

A519944-10G A519944-250MG
76991-450EA 377.69 USD
76991-450 76991-452
2,3-Dibromotoluene 95%
2,3-Dibromotoluene

Precaución: for Research Use Only

Formula: C₇H₆Br₂
MW: 249.9305 g/mol
MDL Number: MFCD11656704
CAS Number: 61563-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR