tert-Butyl-4-(4-amino-2-fluorophenyl)tetrahydro-1(2H)-pyrazinecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-(4-amino-2-fluorophenyl)piperazine-1-carboxylate, 4-(4-Amino-2-fluorophenyl)piperazine, N1-BOC protected

A519489-100MG A519489-1G A519489-250MG A519489-5G A519489-25G
77034-658EA 7 USD
77034-658 77034-660 77034-662 77034-664 77872-870
tert-Butyl-4-(4-amino-2-fluorophenyl)tetrahydro-1(2H)-pyrazinecarboxylate 98%
tert-Butyl-4-(4-amino-2-fluorophenyl)tetrahydro-1(2H)-pyrazinecarboxylate

Precaución: for Research Use Only

Formula: C₁₅H₂₂FN₃O₂
MW: 295.36 g/mol
MDL Number: MFCD08443960
CAS Number: 154590-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR