6-Chloro-1,3-benzoxazol-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-6-chlorobenzoxazole

A519436-1G A519436-250MG
77004-830EA 27.59 USD
77004-830 77004-832
6-Chloro-1,3-benzoxazol-2-amine 97%
6-Chloro-1,3-benzoxazol-2-amine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂O
MW: 168.58 g/mol
MDL Number: MFCD01664218
CAS Number: 52112-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR