6-Chloro-2-fluoro-3-pyridineboronic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(6-Chloro-2-fluoropyridin-3-yl)boronic acid, 6-Chloro-2-methylpyridine-3-boronic acid, 6-Chloro-2-fluoropyridine-3-boronic acid
A518793-250MG
A518793-5G
A518793-1G
76991-446EA
57.29
USD
76991-446
76991-448
77684-347
6-Chloro-2-fluoro-3-pyridineboronic acid 98%
6-Chloro-2-fluoro-3-pyridineboronic acid
Precaución: For research use only.
Formula:
C₅H₄BClFNO₂ MW: 175.35 g/mol |
MDL Number:
MFCD11504861 CAS Number: 1256345-66-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...