8-Chloro-5-nitroquinoline 98%

Supplier: Ambeed
Warning

A518705-100MG A518705-250MG A518705-5G A518705-1G
77034-652EA 24.41 USD
77034-652 77034-654 77034-656 77673-378
8-Chloro-5-nitroquinoline 98%
8-Chloro-5-nitroquinoline

Precaución: for Research Use Only

Formula: C₉H₅ClN₂O₂
MW: 208.6000 g/mol
MDL Number: MFCD00466079
CAS Number: 22539-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR