Methyl-2,3-dimethylbenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,3-dimethylbenzoate

A518470-100G A518470-10G A518470-1G A518470-25G A518470-5G
76991-436EA 627 USD
76991-436 76991-438 76991-440 76991-442 76991-444
Methyl-2,3-dimethylbenzoate 97%
Methyl 2,3-dimethylbenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₂
MW: 164.2000 g/mol
MDL Number: MFCD06203773
CAS Number: 15012-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR