5-Bromo-3-methoxy-1H-pyrazolo[3,4-b]pyridine 95%

Supplier: Ambeed
Warning

A517433-100MG A517433-1G
77017-380EA 62.6 USD
77017-380 77017-382
5-Bromo-3-methoxy-1H-pyrazolo[3,4-b]pyridine 95%
5-Bromo-3-methoxy-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₇H₆BrN₃O
MW: 228.05 g/mol
MDL Number: MFCD22566115
CAS Number: 1363381-82-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR