4-(Benzyloxy)pyridin-2-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-4-benzyloxypyridine

A517002-1G A517002-250MG A517002-25G A517002-5G
77004-822EA 50.93 USD
77004-822 77004-824 77004-826 77004-828
4-(Benzyloxy)pyridin-2-amine 98%
4-(Benzyloxy)pyridin-2-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₂N₂O
MW: 200.2400 g/mol
MDL Number: MFCD12405835
CAS Number: 85333-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR