5-Bromo-2-formyl-3-methoxypyridine 98%

Supplier: Ambeed
Warning

Synonyms: 5-Bromo-3-methoxypicolinaldehyde

A516733-1G A516733-250MG
77004-818EA 189 USD
77004-818 77004-820
5-Bromo-2-formyl-3-methoxypyridine 98%
5-Bromo-2-formyl-3-methoxypyridine

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.0300 g/mol
MDL Number: MFCD11857632
CAS Number: 1087659-24-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR