4-Bromo-2,2-diphenylbutanenitrile 95%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-2,2-diphenylbutyronitrile

A516618-100G A516618-10G A516618-25G A516618-5G
77034-644EA 30.77 USD
77034-644 77034-646 77034-648 77034-650
4-Bromo-2,2-diphenylbutanenitrile 95%
4-Bromo-2,2-diphenylbutanenitrile

Precaución: for Research Use Only

Formula: C₁₆H₁₄BrN
MW: 300.20 g/mol
MDL Number: MFCD00001845
CAS Number: 39186-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR