Methyl 2-chloro-5-methoxybenzoate 98%

Supplier: Ambeed
Warning

A516411-100MG A516411-1G A516411-250MG A516411-5G
77004-808EA 8 USD
77004-808 77004-810 77004-812 77004-814
Methyl 2-chloro-5-methoxybenzoate 98%
Methyl 2-chloro-5-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₉ClO₃
MDL Number: MFCD12913365
CAS Number: 54810-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR