5-Bromo-1H-inden-1-one 95%

Supplier: Ambeed
Warning

A516157-100MG A516157-1G A516157-250MG
76991-420EA 97.61 USD
76991-420 76991-422 76991-424
5-Bromo-1H-inden-1-one 95%
5-Bromo-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₉H₅BrO
MW: 209.03 g/mol
MDL Number: MFCD09842501
CAS Number: 338989-48-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR