[4-(Tetrahydrofurfurylamino-1-carbonyl)phenyl]boronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: N-((Tetrahydrofuran-2-yl)methyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzamide

A515127-1G A515127-250MG A515127-5G
77017-360EA 81 USD
77017-360 77017-362 77017-364
[4-(Tetrahydrofurfurylamino-1-carbonyl)phenyl]boronic acid pinacol ester 97%
[4-(Tetrahydrofurfurylamino-1-carbonyl)phenyl]boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₆BNO₄
MDL Number: MFCD09266200
CAS Number: 1073355-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR