Potassium perfluorobutane-1-sulfonate 98%

Supplier: Ambeed
Warning

Synonyms: Nonafluoro-1-butanesulphonic acid potassium salt, Potassium Perfluoro-1-butanesulphonate, Potassium Nonafluoro-1-butanesulphonate, Perfluoro-1-butanesulphonic acid potassium salt, Potassium nonafluoro-1-butanesulphonate

A514607-10G A514607-500G A514607-5G A514607-100G
77004-796EA 8.49 USD
77004-796 77004-798 77004-800 77594-606
Potassium perfluorobutane-1-sulfonate 98%
Potassium perfluorobutane-1-sulphonate

Precaución: for Research Use Only

Formula: C₄F₉KO₃S
MW: 338.1899 g/mol
MDL Number: MFCD00007515
CAS Number: 29420-49-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR