(S)-(4-(3-Fluoro-5-(trifluoromethyl)pyridin-2-yl)piperazin-1-yl)(5-(methylsulfonyl)-2-((1,1,1-trifluoropropan-2-yl)oxy)phenyl)methanone 98%
Supplier: Ambeed
|
Warning
|
A514249-10MG
A514249-1MG
A514249-2MG
76991-404EA
81
USD
76991-404
76991-406
76991-408
76991-410
(S)-(4-(3-Fluoro-5-(trifluoromethyl)pyridin-2-yl)piperazin-1-yl)(5-(methylsulfonyl)-2-((1,1,1-trifluoropropan-2-yl)oxy)phenyl)methanone 98%
(S)-(4-(3-Fluoro-5-(trifluoromethyl)pyridin-2-yl)piperazin-1-yl)(5-(methylsulfonyl)-2-((1,1,1-trifluoropropan-2-yl)oxy)phenyl)methanone
Precaución: for Research Use Only
Formula:
C₂₁H₂₀F₇N₃O₄S |
MDL Number:
MFCD18251496 CAS Number: 845614-11-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...