6-Chloro-3-methyl-3H-imidazo[4,5-c]pyridine 95%

Supplier: Ambeed
Warning

A514161-250MG A514161-1G
76991-398EA 78 USD
76991-398 77891-037
6-Chloro-3-methyl-3H-imidazo[4,5-c]pyridine 95%
6-Chloro-3-methyl-3H-imidazo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₇H₆ClN₃
MW: 167.59 g/mol
MDL Number: MFCD26743555
CAS Number: 7205-45-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR