1,4-Bis(Boc)-2-piperazinemethanol 97%

Supplier: Ambeed
Warning

A513684-1G A513684-250MG A513684-5G
76991-392EA 155.96 USD
76991-392 76991-394 76991-396
1,4-Bis(Boc)-2-piperazinemethanol 97%
1,4-Bis(Boc)-2-piperazinemethanol

Precaución: for Research Use Only

Formula: C₁₅H₂₈N₂O₅
MW: 316.39 g/mol
MDL Number: MFCD09908093
CAS Number: 143540-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR