(2R,3R,4R,5R)-2-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
DMT-2'O-Methyl-rU Phosphoramidite
A513418-10G
A513418-1G
A513418-5G
A513418-25G
76991-386EA
195.21
USD
76991-386
76991-388
76991-390
77684-338
(2R,3R,4R,5R)-2-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite 95%
(2R,3R,4R,5R)-2-((Bis(4-methoxyphenyl)(phenyl)methoxy)methyl)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-4-methoxytetrahydrofuran-3-yl (2-cyanoethyl) diisopropylphosphoramidite
Precaución: For research use only.
Formula:
C₄₀H₄₉N₄O₉P MW: 760.81 g/mol |
MDL Number:
MFCD00792626 CAS Number: 110764-79-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...