1,4-Diphenylbutadiyne 98%

Supplier: Ambeed
Warning

A513377-100G A513377-10G A513377-1G A513377-25G A513377-5G
77034-622EA 203 USD
77034-622 77034-624 77034-626 77034-628 77034-630
1,4-Diphenylbutadiyne 98%
1,4-Diphenylbutadiyne

Precaución: for Research Use Only

Formula: (C₆H₅)CCCC(C₆H₅)
MW: 202.26
Boiling Pt: 338
Melting Pt: 87
Density: 1.163
MDL Number: MFCD00004787
CAS Number: 886-66-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR