5,5,10,10-Tetramethyl-5,10-dihydroindeno[2,1-a]indene 98%

Supplier: Ambeed
Warning

A513244-100MG A513244-250MG
76991-376EA 55 USD
76991-376 76991-378
5,5,10,10-Tetramethyl-5,10-dihydroindeno[2,1-a]indene 98%
5,5,10,10-Tetramethyl-5,10-dihydroindeno[2,1-a]indene

Precaución: for Research Use Only

Formula: C₂₀H₂₀
MDL Number: MFCD26403658
CAS Number: 89057-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR