6-Bromo-1H-benzo[d]imidazole-2-carbaldehyde 98%

Supplier: Ambeed
Warning

A512833-100MG A512833-250MG
77017-336EA 64.72 USD
77017-336 77017-338
6-Bromo-1H-benzo[d]imidazole-2-carbaldehyde 98%
6-Bromo-1H-benzo[d]imidazole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C₈H₅BrN₂O
MW: 225.04 g/mol
MDL Number: MFCD06738772
CAS Number: 885280-26-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR