4-Fluoro-2-morpholinoaniline 95%

Supplier: Ambeed
Warning

A512682-1G A512682-250MG A512682-5G
77017-328EA 60 USD
77017-328 77017-330 77017-332
4-Fluoro-2-morpholinoaniline 95%
4-Fluoro-2-morpholinoaniline

Precaución: for Research Use Only

Formula: C₁₀H₁₃FN₂O
MDL Number: MFCD11849334
CAS Number: 232951-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR