6-Methyl-1H-pyrrolo[3,2-b]pyridine 98%

Supplier: Ambeed
Warning

A512193-100MG A512193-250MG
76991-370EA 103.97 USD
76991-370 76991-372
6-Methyl-1H-pyrrolo[3,2-b]pyridine 98%
6-Methyl-1H-pyrrolo[3,2-b]pyridine

Precaución: for Research Use Only

Formula: C₈H₈N₂
MW: 132.16 g/mol
MDL Number: MFCD11109592
CAS Number: 1175015-76-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR