tert-Butyl-4-benzoylpiperidine-1-carboxylate 96%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 4-benzoylpiperidine-1-carboxylate

A511976-5G A511976-25G
77034-602EA 230.22 USD
77034-602 77673-356
tert-Butyl-4-benzoylpiperidine-1-carboxylate 96%
tert-Butyl-4-benzoylpiperidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₇H₂₃NO₃
MW: 289.38 g/mol
MDL Number: MFCD09832216
CAS Number: 193217-39-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR