D-(-)-Norleucine 98%

Supplier: Ambeed
Warning

Synonyms: H-D-Nle-OH, (R)-(-)-Norleucine

A511943-100G A511943-1G A511943-25G A511943-5G
77004-774EA 187.78 USD
77004-774 77004-776 77004-778 77004-780
D-(-)-Norleucine 98%
D-(-)-Norleucine

Precaución: for Research Use Only

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
MDL Number: MFCD00008099
CAS Number: 327-56-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR