3-Amino-6-fluoro-2-methylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Fluoro-2-methylpyridin-3-amine

A511857-1G A511857-5G
77004-770EA 25.47 USD
77004-770 77004-772
3-Amino-6-fluoro-2-methylpyridine 98%
3-Amino-6-fluoro-2-methylpyridine

Precaución: for Research Use Only

Formula: C₆H₇FN₂
MW: 126.13 g/mol
MDL Number: MFCD03094949
CAS Number: 28489-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR