5-Bromo-2-fluoro-4-methoxypyridine 98%

Supplier: Ambeed
Warning

A511593-1G A511593-100MG A511593-250MG
77017-320EA 143 USD
77017-320 77613-608 77891-036
5-Bromo-2-fluoro-4-methoxypyridine 98%
5-Bromo-2-fluoro-4-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₅BrFNO
MDL Number: MFCD18255778
CAS Number: 1211588-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR