2-(4-(Dimethylamino)phenyl)-1H-benzo[d]imidazol-5-amine 95%

Supplier: Ambeed
Warning

A511236-100MG A511236-1G A511236-250MG
77004-764EA 66.84 USD
77004-764 77004-766 77004-768
2-(4-(Dimethylamino)phenyl)-1H-benzo[d]imidazol-5-amine 95%
2-(4-(Dimethylamino)phenyl)-1H-benzo[d]imidazol-5-amine

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₄
MW: 252.31 g/mol
MDL Number: MFCD00432727
CAS Number: 40655-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR