4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 98%

Supplier: Ambeed
Warning

A510947-1G A510947-250MG A510947-5G A510947-10G
77034-596EA 24 USD
77034-596 77034-598 77034-600 77594-602
4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid 98%
4-(N-Maleimidomethyl)cyclohexane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅NO₄
MDL Number: MFCD03093740
CAS Number: 64987-82-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR