(1S,2S)-1-Amino-2-(diphenylphosphino)indane 98%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-2-(Diphenylphosphino)-2,3-dihydro-1H-inden-1-amine

A510913-100MG A510913-1G A510913-250MG
76991-360EA 14 USD
76991-360 76991-362 76991-364
(1S,2S)-1-Amino-2-(diphenylphosphino)indane 98%
(1S,2S)-1-Amino-2-(diphenylphosphino)indane

Precaución: for Research Use Only

Formula: C₂₁H₂₀NP
MDL Number: MFCD17013984
CAS Number: 1091606-69-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR