4-Bromo-3,5-dimethylpyridine hydrochloride 95%

Supplier: Ambeed
Warning

A510687-100MG A510687-250MG
77017-310EA 78 USD
77017-310 77017-312
4-Bromo-3,5-dimethylpyridine hydrochloride 95%
4-Bromo-3,5-dimethylpyridine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈BrN·ClH
MDL Number: MFCD12922840
CAS Number: 201286-65-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR