Methyl 3-hydroxycyclohexanecarboxylate 95%

Supplier: Ambeed
Warning

A510561-100MG A510561-1G A510561-250MG A510561-5G
76991-350EA 10 USD
76991-350 76991-352 76991-354 76991-356
Methyl 3-hydroxycyclohexanecarboxylate 95%
Methyl 3-hydroxycyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₄O₃
MDL Number: MFCD11975826
CAS Number: 37722-82-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR