4-(1H-Pyrazol-3-yl)benzoic acid 95%

Supplier: Ambeed
Warning

Synonyms: 3-(4-Carboxyphenyl)-1H-pyrazole

A510273-100MG A510273-1G A510273-250MG A510273-5G
76991-342EA 31.83 USD
76991-342 76991-344 76991-346 76991-348
4-(1H-Pyrazol-3-yl)benzoic acid 95%
4-(1H-Pyrazol-3-yl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.183 g/mol
MDL Number: MFCD01569442
CAS Number: 208511-67-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR