BippyPhos 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-(DI-T-BUTYLPHOSPHINO)-1',3',5'-TRIPHENYL-1,4'-BI-1H-PYRAZOLE, 5-(Di-tert-butylphosphino)-1', 3', 5'-triphenyl-1'H-[1,4']bipyrazole, 5-(di-tert-Butylphosphino)-1',3',5'-triphenyl-1'H-1,4'-bipyrazole, 5-(Di-tert-butylphosphino)-1',3',5'-triphenyl-1'H-1,4-bipyrazole
A510069-10G
A510069-1G
A510069-250MG
A510069-25G
A510069-5G
A510069-100G
77034-570EA
54
USD
77034-570
77034-572
77034-574
77034-576
77034-578
77602-182
BippyPhos 98%
BippyPhos
Precaución: for Research Use Only
Formula:
C₃₂H₃₅N₄P MW: 506.634 g/mol |
MDL Number:
MFCD09038440 CAS Number: 894086-00-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...