2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzoxazole 95%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[d]oxazole

A509996-100MG A509996-1G A509996-250MG A509996-5G
77017-296EA 58.35 USD
77017-296 77017-298 77017-300 77017-302
2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzoxazole 95%
2-Methyl-6-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3-benzoxazole

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₃
MW: 259.109 g/mol
MDL Number: MFCD13181971
CAS Number: 1408089-23-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR