(4-Ethoxy-2,5-dimethylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A508821-1G A508821-250MG A508821-5G
77034-558EA 35.01 USD
77034-558 77034-560 77034-562
(4-Ethoxy-2,5-dimethylphenyl)boronic acid 98%
(4-Ethoxy-2,5-dimethylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.03 g/mol
MDL Number: MFCD12026704
CAS Number: 1217500-56-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR