2-Amino-3-chloro-6-methoxypyridine 98%

Supplier: Ambeed
Warning

A508564-1G A508564-250MG A508564-5G A508564-100MG
77004-746EA 71.09 USD
77004-746 77004-748 77004-750 77673-336
2-Amino-3-chloro-6-methoxypyridine 98%
2-Amino-3-chloro-6-methoxypyridine

Precaución: for Research Use Only

Formula: C₆H₇ClN₂O
MW: 158.586 g/mol
MDL Number: MFCD11044247
CAS Number: 742070-73-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR