(4-(2-Oxo-2-(propylamino)ethyl)phenyl)boronic acid 96%

Supplier: Ambeed
Warning

A508214-100MG A508214-1G A508214-250MG
77017-278EA 39 USD
77017-278 77017-280 77017-282
(4-(2-Oxo-2-(propylamino)ethyl)phenyl)boronic acid 96%
(4-(2-Oxo-2-(propylamino)ethyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₆BNO₃
MDL Number: MFCD17015734
CAS Number: 1256345-81-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR