[(4-Fluorophenyl)amino](oxo)acetic acid 98%

Supplier: Ambeed
Danger

A508185-25G A508185-5G A508185-1G
77034-550EA 269 USD
77034-550 77034-552 77872-844
[(4-Fluorophenyl)amino](oxo)acetic acid 98%
[(4-Fluorophenyl)amino](oxo)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆FNO₃
MDL Number: MFCD09864524
CAS Number: 69066-43-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR