1-(4-Chlorophenyl)-1H-pyrazole-4-carbaldehyde 97%

Supplier: Ambeed
Warning

A507606-1G A507606-250MG
77034-548EA 106.09 USD
77034-548 77673-332
1-(4-Chlorophenyl)-1H-pyrazole-4-carbaldehyde 97%
1-(4-Chlorophenyl)-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₇ClN₂O
MW: 206.63 g/mol
MDL Number: MFCD07643175
CAS Number: 63874-99-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR