Sodium Pyridine-2-sulfinate 95%

Supplier: Ambeed
Warning

A507155-5G A507155-100MG A507155-250MG
77017-274EA 947.99 USD
77017-274 77891-029 77891-030
Sodium Pyridine-2-sulfinate 95%
Sodium Pyridine-2-sulfinate

Precaución: for Research Use Only

Formula: C₅H₄NNaO₂S
MDL Number: MFCD20483664
CAS Number: 24367-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR