4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine 96%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl [4,4'-bipiperidine]-1-carboxylate, 4,4'-Bipiperidine, 1-BOC protected, N-Boc-4,4'-bipiperidine

A507030-100MG A507030-1G A507030-250MG A507030-5G
77017-266EA 35.01 USD
77017-266 77017-268 77017-270 77017-272
4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine 96%
4-(4'-Piperid-1-yl)-1-tert-butoxycarbonylpiperidine

Precaución: for Research Use Only

Formula: C₁₅H₂₈N₂O₂
MW: 268.4 g/mol
MDL Number: MFCD02179169
CAS Number: 171049-35-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR